Assay Details
| Gene Symbol: Yme1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013771.5 | NP_038799.1 | 4 - 5 | 617 | 92 | |
| GenBank mRNA | AF090430.1 | - | 4 - 5 | 595 | 92 | |
| AK032273.1 | - | 4 - 5 | 605 | 92 | ||
| AK045141.1 | - | 4 - 5 | 602 | 92 | ||
| AK079175.1 | - | 4 - 5 | 577 | 92 | ||
| AK137665.1 | - | 4 - 5 | 569 | 92 | ||
| AY136286.1 | - | 4 - 5 | 433 | 92 | ||
| AY136287.1 | - | 4 - 5 | 586 | 92 | ||
| BC007128.1 | - | 4 - 5 | 578 | 3591123 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yme1l1
Genomic Map