Assay Details
| Gene Symbol: Yme1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013771.5 | NP_038799.1 | 8 - 9 | 1050 | 84 | |
| GenBank mRNA | AF090430.1 | - | 8 - 9 | 1028 | 84 | |
| AK045141.1 | - | 8 - 9 | 1035 | 84 | ||
| AK079175.1 | - | 8 - 9 | 1010 | 84 | ||
| AK137665.1 | - | 8 - 9 | 1002 | 84 | ||
| AY136286.1 | - | 8 - 9 | 866 | 84 | ||
| AY136287.1 | - | 8 - 9 | 1019 | 84 | ||
| BC007128.1 | - | 8 - 9 | 1011 | 3591123 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yme1l1
Genomic Map