Assay Details
| Gene Symbol: Myh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177369.3 | NP_796343.2 | 19 - 20 | 2353 | 70 | |
| GenBank mRNA | AK029328.1 | - | 19 - 20 | 2264 | 70 | |
| BC150737.1 | - | 19 - 20 | 2276 | 9087648 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myh8
Genomic Map