Assay Details
| Gene Symbol: Lin7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015399.1 | - | 1 - 2 | 64 | 102 | |
| AK078923.1 | - | 1 - 2 | 65 | 102 | ||
| AK083299.1 | - | 1 - 2 | 68 | 102 | ||
| AK128989.1 | - | 1 - 2 | 65 | 102 | ||
| AK135810.1 | - | 1 - 2 | 71 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015399.1, AK078923.1, AK135810.1 transcripts.
Set Membership: |
|

Lin7c
Genomic Map