Assay Details
| Gene Symbol: Ptpra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163688.1 | NP_001157160.1 | 13 - 14 | 1721 | 79 | |
| NM_008980.2 | NP_033006.2 | 14 - 15 | 1829 | 79 | ||
| GenBank mRNA | AF332081.1 | - | 12 - 13 | 1434 | 79 | |
| AF332082.1 | - | 12 - 13 | 1434 | 79 | ||
| AJ431367.1 | - | 12 - 13 | 1434 | 79 | ||
| AK031690.1 | - | 13 - 14 | 1720 | 79 | ||
| AK165673.1 | - | 13 - 14 | 1719 | 79 | ||
| BC050930.1 | - | 13 - 14 | 1703 | 6811172 | 79 | |
| BC110683.1 | - | 13 - 14 | 1687 | 3965311 | 79 | |
| M36033.1 | - | 14 - 15 | 1567 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpra
Genomic Map