Assay Details
| Gene Symbol: Slc4a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081162.1 | NP_001074631.1 | 17 - 18 | 2265 | 67 | |
| GenBank mRNA | AK141286.1 | - | 3 - 4 | 524 | 67 | |
| AK158115.1 | - | 17 - 18 | 68 | |||
| BC111884.1 | - | 16 - 17 | 2060 | 40060299 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a11
Genomic Map