Assay Details
| Gene Symbol: Slc4a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081162.1 | NP_001074631.1 | 18 - 19 | 2436 | 64 | |
| GenBank mRNA | AK141286.1 | - | 4 - 5 | 695 | 64 | |
| AK158115.1 | - | 18 - 19 | 2391 | 64 | ||
| BC036154.1 | - | 7 - 8 | 2104 | 64 | ||
| BC111884.1 | - | 17 - 18 | 2231 | 40060299 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a11
Genomic Map