Assay Details
| Gene Symbol: Slc4a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081162.1 | NP_001074631.1 | 19 - 20 | 2630 | 86 | |
| GenBank mRNA | AK141286.1 | - | 5 - 6 | 889 | 86 | |
| AK158115.1 | - | 19 - 20 | 2585 | 86 | ||
| BC036154.1 | - | 8 - 9 | 2298 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a11
Genomic Map