Assay Details
| Gene Symbol: Slc4a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081162.1 | NP_001074631.1 | 2 - 3 | 270 | 63 | |
| GenBank mRNA | AK052665.1 | - | 2 - 3 | 221 | 63 | |
| AK158115.1 | - | 2 - 3 | 224 | 63 | ||
| BC111884.1 | - | 1 - 2 | 65 | 40060299 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111884.1, NM_001081162.1 transcripts.
Set Membership: |
|

Slc4a11
Genomic Map