Assay Details
| Gene Symbol: Slc4a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081162.1 | NP_001074631.1 | 4 - 5 | 496 | 112 | |
| GenBank mRNA | AK158115.1 | - | 4 - 5 | 450 | 112 | |
| BC111884.1 | - | 3 - 4 | 291 | 40060299 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a11
Genomic Map