Assay Details
| Gene Symbol: Slc4a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081162.1 | NP_001074631.1 | 8 - 9 | 981 | 71 | |
| GenBank mRNA | AK052665.1 | - | 4 - 5 | 1777 | 71 | |
| AK158115.1 | - | 8 - 9 | 935 | 71 | ||
| BC111884.1 | - | 7 - 8 | 776 | 40060299 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
sodium ion transport
anion transport
bicarbonate transport
proton transport
cellular cation homeostasis
borate transmembrane transport
sodium ion transmembrane transport
fluid transport
borate transport
ion homeostasis
regulation of intracellular pH
transporter activity
sodium channel activity
inorganic anion exchanger activity
anion transmembrane transporter activity
bicarbonate transmembrane transporter activity
hydrogen ion channel activity
symporter activity
anion:anion antiporter activity
borate transmembrane transporter activity
protein dimerization activity
transport
ion transport
sodium ion transport
anion transport
bicarbonate transport
proton transport
cellular cation homeostasis
borate transmembrane transport
sodium ion transmembrane transport
fluid transport
borate transport
ion homeostasis
regulation of intracellular pH
transporter activity
sodium channel activity
inorganic anion exchanger activity
anion transmembrane transporter activity
bicarbonate transmembrane transporter activity
hydrogen ion channel activity
symporter activity
anion:anion antiporter activity
borate transmembrane transporter activity
protein dimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121976 | BC036154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121976 | AK158115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121976 | AK141286.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121976 | NM_001081162.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20363 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731308 | AK017784.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326576 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244295 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 23429 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108634 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139600 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121976 | BC036154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121976 | AK158115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121976 | AK141286.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121976 | NM_001081162.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00493366 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04692870 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139600 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21631162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01577626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01618976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02110923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02106216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01772559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01593628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01577627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01577625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02110921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01593623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01577624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01887498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01976480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01618980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01432701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02048837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02082875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02106215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02110905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02048832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01772557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21631278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01432698 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10487736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487740 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487743 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487723 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487726 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487729 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487734 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487733 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487728 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487747 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487744 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487739 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487741 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487745 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487731 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487732 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487727 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457989_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434867_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10487722 | - | - |
| Mouse Genome 430 2.0 Array | 1457989_at | - | - |
| Mouse Genome 430 2.0 Array | 1457989_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434867_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434867_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269356_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269356_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200039678.mm.1 | NM_001081162.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200039678.mm.1 | NM_001081162.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200039678.mm.1 | TC0200004636.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200039680.mm.1 | NM_001081162.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200039680.mm.1 | NM_001081162.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200039680.mm.1 | TC0200004636.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200039682.mm.1 | NM_001081162.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200039682.mm.1 | NM_001081162.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200039682.mm.1 | TC0200004636.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1457989_at | - | - |
| Mouse Expression Set 430 | 1457989_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434867_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457989_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434867_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457989_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 439562 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103830154 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101740056 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2484420 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252747 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255284 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101740056 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101740056 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2484420 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255284 | - | - |

Slc4a11
Genomic Map