Assay Details
| Gene Symbol: Lzts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291027.1 | NP_001277956.1 | 1 - 2 | 126 | 57 | |
| NM_001291028.1 | NP_001277957.1 | 1 - 2 | 126 | 57 | ||
| NM_197945.4 | NP_922936.3 | 1 - 2 | 126 | 57 | ||
| GenBank mRNA | AK148251.1 | - | 1 - 2 | 85 | 57 | |
| AK161711.1 | - | 1 - 2 | 80 | 57 | ||
| DN175861.1 | - | 1 - 2 | 75 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK161711.1, NM_001291027.1, NM_001291028.1, NM_197945.4 transcripts.
Set Membership: |
|

Lzts3
Genomic Map