Assay Details
| Gene Symbol: Lzts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291027.1 | NP_001277956.1 | 3 - 4 | 760 | 77 | |
| NM_197945.4 | NP_922936.3 | 3 - 4 | 760 | 77 | ||
| GenBank mRNA | AK148251.1 | - | 3 - 4 | 719 | 77 | |
| AK161711.1 | - | 3 - 4 | 714 | 77 | ||
| BC049892.1 | - | 1 - 2 | 321 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lzts3
Genomic Map