Assay Details
| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111075.4 | NP_001104545.1 | 10 - 11 | 1333 | 75 | |
| NM_023117.4 | NP_075606.1 | 11 - 12 | 1411 | 75 | ||
| GenBank mRNA | AK004836.1 | - | 10 - 11 | 1290 | 75 | |
| AK043730.1 | - | 10 - 11 | 1324 | 75 | ||
| AK153903.1 | - | 11 - 12 | 1412 | 75 | ||
| BC002287.1 | - | 2 - 3 | 198 | 75 | ||
| BC048164.1 | - | 10 - 11 | 1330 | 75 | ||
| BC057568.1 | - | 10 - 11 | 1204 | 6401489 | 75 | |
| S93521.1 | - | 11 - 12 | 1280 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc25b
Genomic Map