Assay Details
| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111075.4 | NP_001104545.1 | 4 - 5 | 652 | 84 | |
| NM_023117.4 | NP_075606.1 | 4 - 5 | 652 | 84 | ||
| GenBank mRNA | AK004836.1 | - | 4 - 5 | 609 | 84 | |
| AK043730.1 | - | 4 - 5 | 643 | 84 | ||
| AK153903.1 | - | 4 - 5 | 653 | 84 | ||
| AK157397.1 | - | 4 - 5 | 610 | 84 | ||
| BC048164.1 | - | 4 - 5 | 649 | 84 | ||
| BC057568.1 | - | 4 - 5 | 523 | 6401489 | 84 | |
| BQ561430.1 | - | 4 - 5 | 308 | 84 | ||
| S93521.1 | - | 4 - 5 | 521 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc25b
Genomic Map