Assay Details
| Gene Symbol: Nr1h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177730.1 | NP_001171201.1 | 8 - 9 | 1287 | 74 | |
| NM_013839.4 | NP_038867.2 | 8 - 9 | 1440 | 74 | ||
| GenBank mRNA | AF085745.1 | - | 8 - 9 | 1182 | 74 | |
| AJ132601.1 | - | 8 - 9 | 1218 | 74 | ||
| AK143904.1 | - | 8 - 9 | 1221 | 74 | ||
| AK159250.1 | - | 8 - 9 | 1441 | 74 | ||
| AY195870.1 | - | 8 - 9 | 1287 | 74 | ||
| AY195871.1 | - | 8 - 9 | 1287 | 74 | ||
| BC012646.1 | - | 8 - 9 | 1228 | 4163109 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nr1h3
Genomic Map