Assay Details
| Gene Symbol: Slc39a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290765.1 | NP_001277694.1 | 7 - 8 | 989 | 59 | |
| NM_026721.3 | NP_080997.1 | 7 - 8 | 950 | 59 | ||
| NR_110981.1 | 5 - 6 | 553 | 59 | |||
| GenBank mRNA | AK003191.1 | - | 3 - 4 | 252 | 59 | |
| AK035296.1 | - | 7 - 8 | 857 | 59 | ||
| AK156951.1 | - | 7 - 8 | 875 | 59 | ||
| BC020106.1 | - | 5 - 6 | 468 | 59 | ||
| BC100463.1 | - | 7 - 8 | 957 | 30918840 | 59 | |
| BC125525.1 | - | 7 - 8 | 811 | 40130040 | 59 | |
| BC137752.1 | - | 7 - 8 | 811 | 8860771 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc39a13
Genomic Map