Assay Details
| Gene Symbol: Epb4.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013513.2 | NP_038541.1 | 10 - 11 | 1780 | 118 | |
| GenBank mRNA | AK089080.1 | - | 4 - 5 | 828 | 118 | |
| AK133896.1 | - | 10 - 11 | 1794 | 118 | ||
| AK143841.1 | - | 10 - 11 | 1760 | 118 | ||
| AK158219.1 | - | 10 - 11 | 1698 | 118 | ||
| AK158247.1 | - | 10 - 11 | 1744 | 118 | ||
| AK165288.1 | - | 10 - 11 | 1707 | 118 | ||
| AK172410.1 | - | 10 - 11 | 1744 | 118 | ||
| BC053739.1 | - | 10 - 11 | 1752 | 118 | ||
| BC066194.1 | - | 10 - 11 | 1780 | 30429971 | 118 | |
| BC066806.1 | - | 10 - 11 | 1780 | 30429961 | 118 | |
| L35933.1 | - | 10 - 11 | 1759 | 118 | ||
| U03487.1 | - | 10 - 11 | 1710 | 118 | ||
| U04055.1 | - | 10 - 11 | 1780 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epb4.2
Genomic Map