Assay Details
| Gene Symbol: Epb4.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013513.2 | NP_038541.1 | 7 - 8 | 1134 | 105 | |
| GenBank mRNA | AK089080.1 | - | 1 - 2 | 182 | 105 | |
| AK133896.1 | - | 7 - 8 | 1148 | 105 | ||
| AK137484.1 | - | 7 - 8 | 1215 | 105 | ||
| AK143841.1 | - | 7 - 8 | 1114 | 105 | ||
| AK158219.1 | - | 7 - 8 | 1052 | 105 | ||
| AK158247.1 | - | 7 - 8 | 1098 | 105 | ||
| AK158370.1 | - | 7 - 8 | 1061 | 105 | ||
| AK165288.1 | - | 7 - 8 | 1061 | 105 | ||
| AK172410.1 | - | 7 - 8 | 1098 | 105 | ||
| BC066194.1 | - | 7 - 8 | 1134 | 30429971 | 105 | |
| BC066806.1 | - | 7 - 8 | 1134 | 30429961 | 105 | |
| L35933.1 | - | 7 - 8 | 1113 | 105 | ||
| U03487.1 | - | 7 - 8 | 1064 | 105 | ||
| U04055.1 | - | 7 - 8 | 1134 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epb4.2
Genomic Map