Assay Details
| Gene Symbol: Adal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290811.1 | NP_001277740.1 | 1 - 2 | 236 | 116 | |
| GenBank mRNA | AK016299.1 | - | 1 - 2 | 233 | 116 | |
| AK048809.1 | - | 1 - 2 | 237 | 116 | ||
| AK049697.1 | - | 1 - 2 | 229 | 116 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016299.1, AK048809.1, AK049697.1, NM_001290811.1 transcripts.
Set Membership: |
|

Adal
Genomic Map