Assay Details
| Gene Symbol: Adal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290811.1 | NP_001277740.1 | 5 - 6 | 683 | 88 | |
| NM_029475.2 | NP_083751.1 | 5 - 6 | 575 | 88 | ||
| GenBank mRNA | AK016299.1 | - | 5 - 6 | 680 | 88 | |
| AK049697.1 | - | 5 - 6 | 676 | 88 | ||
| BC050879.1 | - | 5 - 6 | 527 | 5687835 | 88 | |
| BC052048.1 | - | 5 - 6 | 527 | 5687835 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adal
Genomic Map