Assay Details
| Gene Symbol: Itpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172584.3 | NP_766172.1 | 7 - 8 | 725 | 74 | |
| GenBank mRNA | AK038913.1 | - | 7 - 8 | 726 | 74 | |
| BC056464.1 | - | 7 - 8 | 706 | 5707377 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itpk1
Genomic Map