Assay Details
| Gene Symbol: Golga5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199004.1 | NP_001185933.1 | 6 - 7 | 1569 | 82 | |
| NM_013747.4 | NP_038775.1 | 6 - 7 | 1411 | 82 | ||
| GenBank mRNA | AB016784.1 | - | 6 - 7 | 1570 | 82 | |
| AF026274.1 | - | 6 - 7 | 1401 | 82 | ||
| AK007592.1 | - | 6 - 7 | 1413 | 82 | ||
| AK138455.1 | - | 6 - 7 | 1570 | 82 | ||
| AK152533.1 | - | 6 - 7 | 1419 | 82 | ||
| AK153010.1 | - | 6 - 7 | 1419 | 82 | ||
| AK168765.1 | - | 6 - 7 | 1415 | 82 | ||
| BC016883.1 | - | 6 - 7 | 2139 | 4235952 | 82 | |
| BC086782.1 | - | 6 - 7 | 1411 | 6813878 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Golga5
Genomic Map