Assay Details
| Gene Symbol: Golga5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199004.1 | NP_001185933.1 | 9 - 10 | 1980 | 103 | |
| NM_013747.4 | NP_038775.1 | 9 - 10 | 1822 | 103 | ||
| GenBank mRNA | AB016784.1 | - | 9 - 10 | 1981 | 103 | |
| AF026274.1 | - | 9 - 10 | 1812 | 103 | ||
| AK138455.1 | - | 9 - 10 | 1981 | 103 | ||
| AK152533.1 | - | 9 - 10 | 1830 | 103 | ||
| AK153010.1 | - | 9 - 10 | 1830 | 103 | ||
| AK168765.1 | - | 9 - 10 | 1826 | 103 | ||
| BC016883.1 | - | 9 - 10 | 2550 | 4235952 | 103 | |
| BC086782.1 | - | 9 - 10 | 1822 | 6813878 | 103 | |
More Information
Set Membership: |
|

Golga5
Genomic Map