Assay Details
| Gene Symbol: Olfr368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146374.1 | NP_666486.1 | 1 | 616 | 73 | |
| GenBank mRNA | BC104308.1 | - | 1 | 616 | 40044244 | 73 |
| BC104309.1 | - | 1 | 616 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr368
Genomic Map