Assay Details
| Gene Symbol: Gm13219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035738.1 | - | 3 - 4 | 323 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700061F12 gene | 1700061F12Rik | -236.8kb | Sense | Chr. 2 - 9189515 - 9197484 on NCBI Build GRCm38 | |
| Downstream | GATA binding protein 3 | Gata3 |
Gata-3, jal |
411.8kb | Anti-sense | Chr. 2 - 9857078 - 9887950 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm13219
Genomic Map