| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200028284.mm.1 |
NM_001113569.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028284.mm.1 |
NM_001113569.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028284.mm.1 |
NM_009295.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028284.mm.1 |
NM_009295.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028284.mm.1 |
TC0200003313.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200028285.mm.1 |
NM_001113569.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028285.mm.1 |
NM_001113569.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028285.mm.1 |
NM_009295.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028285.mm.1 |
NM_009295.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028285.mm.1 |
TC0200003313.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200028286.mm.1 |
NM_001113569.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028286.mm.1 |
NM_001113569.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028286.mm.1 |
NM_009295.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028286.mm.1 |
NM_009295.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028286.mm.1 |
TC0200003313.mm.1 |
MTA TC |