Assay Details
| Gene Symbol: Fpgs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010236.2 | NP_034366.2 | 12 - 13 | 1263 | 66 | |
| GenBank mRNA | AK041952.1 | - | 10 - 11 | 1405 | 66 | |
| AK157040.1 | - | 12 - 13 | 1224 | 66 | ||
| BC005484.1 | - | 12 - 13 | 1206 | 3485401 | 66 | |
| BM944126.1 | - | 3 - 4 | 262 | 66 | ||
| U32197.1 | - | 12 - 13 | 1220 | 66 | ||
| U33557.1 | - | 12 - 13 | 1240 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fpgs
Genomic Map