Assay Details
| Gene Symbol: Fpgs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010236.2 | NP_034366.2 | 3 - 4 | 372 | 84 | |
| GenBank mRNA | AK041952.1 | - | 3 - 4 | 332 | 84 | |
| AK157040.1 | - | 3 - 4 | 333 | 84 | ||
| BC005484.1 | - | 3 - 4 | 315 | 3485401 | 84 | |
| CA558410.1 | - | 3 - 4 | 225 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fpgs
Genomic Map