Assay Details
| Gene Symbol: Fpgs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010236.2 | NP_034366.2 | 7 - 8 | 694 | 61 | |
| GenBank mRNA | AK157040.1 | - | 7 - 8 | 655 | 61 | |
| BC005484.1 | - | 7 - 8 | 637 | 3485401 | 61 | |
| CF749232.1 | - | 4 - 5 | 293 | 61 | ||
| U32197.1 | - | 7 - 8 | 651 | 61 | ||
| U33557.1 | - | 7 - 8 | 671 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fpgs
Genomic Map