Assay Details
| Gene Symbol: Fam129b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146119.2 | NP_666231.1 | 6 - 7 | 922 | 62 | |
| GenBank mRNA | AK033735.1 | - | 6 - 7 | 890 | 62 | |
| AK046583.1 | - | 6 - 7 | 819 | 62 | ||
| AK051396.1 | - | 6 - 7 | 861 | 62 | ||
| AK146953.1 | - | 6 - 7 | 907 | 62 | ||
| AK149888.1 | - | 6 - 7 | 832 | 62 | ||
| AK150642.1 | - | 6 - 7 | 860 | 62 | ||
| AK152207.1 | - | 6 - 7 | 881 | 62 | ||
| AK153489.1 | - | 6 - 7 | 866 | 62 | ||
| AK170512.1 | - | 6 - 7 | 862 | 62 | ||
| AK185988.1 | - | 1 - 2 | 58 | 62 | ||
| BC027843.1 | - | 6 - 7 | 842 | 5343013 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam129b
Genomic Map