Assay Details
| Gene Symbol: Gnptg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172529.3 | NP_766117.2 | 1 - 2 | 55 | 70 | |
| GenBank mRNA | AK044021.1 | - | 1 - 2 | 64 | 70 | |
| AK078230.1 | - | 63 | 70 | |||
| BC023869.1 | - | 1 - 2 | 67 | 70 | ||
| BC055872.1 | - | 1 - 2 | 55 | 4925332 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnptg
Genomic Map