Assay Details
| Gene Symbol: Gnptg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172529.3 | NP_766117.2 | 2 - 3 | 117 | 79 | |
| GenBank mRNA | AK078230.1 | - | 125 | 79 | ||
| AY465529.1 | - | 2 - 3 | 125 | 79 | ||
| BC055872.1 | - | 2 - 3 | 117 | 4925332 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnptg
Genomic Map