Assay Details
| Gene Symbol: Tgm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC116266.1 | - | 3 - 4 | 526 | 40094000 | 112 |
| BC116267.1 | - | 3 - 4 | 526 | 40094002 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tgm5
Genomic Map