Assay Details
| Gene Symbol: Tgm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC116266.1 | - | 4 - 5 | 639 | 40094000 | 92 |
| BC116267.1 | - | 4 - 5 | 639 | 40094002 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tgm5
Genomic Map