Assay Details
| Gene Symbol: Tgm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028799.2 | NP_083075.1 | 6 - 7 | 960 | 90 | |
| GenBank mRNA | AK009196.2 | - | 6 - 7 | 960 | 90 | |
| AK132482.1 | - | 6 - 7 | 1043 | 90 | ||
| BC116266.1 | - | 6 - 7 | 943 | 40094000 | 90 | |
| BC116267.1 | - | 6 - 7 | 943 | 40094002 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tgm5
Genomic Map