Assay Details
| Gene Symbol: Tgm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028799.2 | NP_083075.1 | 8 - 9 | 1210 | 78 | |
| GenBank mRNA | AK009196.2 | - | 8 - 9 | 1210 | 78 | |
| AK037074.1 | - | 8 - 9 | 1271 | 78 | ||
| AK132482.1 | - | 8 - 9 | 1293 | 78 | ||
| BC116266.1 | - | 8 - 9 | 1193 | 40094000 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tgm5
Genomic Map