Assay Details
| Gene Symbol: Zfp106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011743.2 | NP_035873.2 | 1 - 2 | 145 | 61 | |
| GenBank mRNA | AK089359.1 | - | 1 - 2 | 143 | 61 | |
| AK147887.1 | - | 1 - 2 | 146 | 61 | ||
| AK155019.1 | - | 1 - 2 | 143 | 61 | ||
| BC025424.1 | - | 1 - 2 | 137 | 4459713 | 61 | |
| CJ104804.1 | - | 1 - 2 | 146 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK089359.1, AK155019.1, BC025424.1, NM_011743.2 transcripts.
Set Membership: |
|

Zfp106
Genomic Map