Assay Details
| Gene Symbol: Zfp106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011743.2 | NP_035873.2 | 2 - 3 | 232 | 82 | |
| GenBank mRNA | AK089359.1 | - | 2 - 3 | 230 | 82 | |
| AK147887.1 | - | 2 - 3 | 233 | 82 | ||
| AK155019.1 | - | 2 - 3 | 230 | 82 | ||
| BC010440.1 | - | 1 - 2 | 72 | 82 | ||
| CJ104804.1 | - | 2 - 3 | 233 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp106
Genomic Map