Assay Details
| Gene Symbol: Zfp106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011743.2 | NP_035873.2 | 3 - 4 | 292 | 60 | |
| GenBank mRNA | AK089359.1 | - | 3 - 4 | 290 | 60 | |
| AK147887.1 | - | 3 - 4 | 293 | 60 | ||
| AK155019.1 | - | 3 - 4 | 290 | 60 | ||
| BC025424.1 | - | 3 - 4 | 281 | 4459713 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp106
Genomic Map