Assay Details
| Gene Symbol: Phpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029293.2 | NP_083569.1 | 2 - 3 | 318 | 117 | |
| GenBank mRNA | AK005756.1 | - | 2 - 3 | 318 | 117 | |
| AK154580.1 | - | 2 - 3 | 338 | 117 | ||
| BC028657.1 | - | 2 - 3 | 291 | 1246740 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
positive regulation of insulin secretion involved in cellular response to glucose stimulus
peptidyl-histidine dephosphorylation
negative regulation of T cell receptor signaling pathway
negative regulation of lyase activity
positive regulation of cell motility
regulation of actin cytoskeleton reorganization
negative regulation of ATP citrate synthase activity
phosphoprotein phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
protein tyrosine/threonine phosphatase activity
JUN kinase phosphatase activity
phosphohistidine phosphatase activity
inositol bisphosphate phosphatase activity
hydrolase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
inositol-1,3,4-trisphosphate 4-phosphatase activity
NADP phosphatase activity
transmembrane receptor protein phosphatase activity
calcium channel inhibitor activity
inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
inositol-1,4,5,6-tetrakisphosphate 6-phosphatase activity
inositol-4,5-bisphosphate 5-phosphatase activity
5-amino-6-(5-phosphoribitylamino)uracil phosphatase activity
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
ion channel binding
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
inositol phosphate phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 1-phosphatase activity
inositol-3,4-bisphosphate 4-phosphatase activity
inositol-1,3,4-trisphosphate 1-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 6-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 1-phosphatase activity
phosphatidylinositol-1,4,5-trisphosphate 5-phosphatase activity
IDP phosphatase activity
protein dephosphorylation
positive regulation of insulin secretion involved in cellular response to glucose stimulus
peptidyl-histidine dephosphorylation
negative regulation of T cell receptor signaling pathway
negative regulation of lyase activity
positive regulation of cell motility
regulation of actin cytoskeleton reorganization
negative regulation of ATP citrate synthase activity
phosphoprotein phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
protein tyrosine/threonine phosphatase activity
JUN kinase phosphatase activity
phosphohistidine phosphatase activity
inositol bisphosphate phosphatase activity
hydrolase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
inositol-1,3,4-trisphosphate 4-phosphatase activity
NADP phosphatase activity
transmembrane receptor protein phosphatase activity
calcium channel inhibitor activity
inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
inositol-1,4,5,6-tetrakisphosphate 6-phosphatase activity
inositol-4,5-bisphosphate 5-phosphatase activity
5-amino-6-(5-phosphoribitylamino)uracil phosphatase activity
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
ion channel binding
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
inositol phosphate phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 1-phosphatase activity
inositol-3,4-bisphosphate 4-phosphatase activity
inositol-1,3,4-trisphosphate 1-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 6-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 1-phosphatase activity
phosphatidylinositol-1,4,5-trisphosphate 5-phosphatase activity
IDP phosphatase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073068 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073067 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073069 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073070 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073066 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P184910 | NM_029293.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P184910 | AK005756.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P184910 | AK154580.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P184910 | BC028657.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P184910 | NM_029293.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P184910 | AK005756.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P184910 | AK154580.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P184910 | BC028657.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38539 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427631 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134245 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P184910 | NM_029293.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P184910 | AK005756.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P184910 | AK154580.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P184910 | BC028657.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P184910 | NM_029293.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P184910 | AK005756.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P184910 | AK154580.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P184910 | BC028657.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073053 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134245 | - | - |
| Whole Mouse Genome G4122A | A_51_P184910 | NM_029293.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P184910 | AK005756.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P184910 | AK154580.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P184910 | BC028657.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P184910 | NM_029293.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P184910 | AK005756.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P184910 | AK154580.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P184910 | BC028657.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134245 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01344177 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01344176 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01344184 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01344172 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073068 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073070 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073066 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073067 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21073069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073061 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10480807 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480805 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480806 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427903_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10480804 | - | - |
| Mouse Genome 430 2.0 Array | 1427903_at | - | - |
| Mouse Genome 430 2.0 Array | 1427903_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75454_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75454_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200026168.mm.1 | NM_029293.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200026168.mm.1 | NM_029293.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200026168.mm.1 | TC0200005494.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200046683.mm.1 | NM_029293.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200046683.mm.1 | NM_029293.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200046683.mm.1 | TC0200005494.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427903_PM_at | - | - |
| Mouse Expression Set 430 | 1427903_at | - | - |
| Murine Genome U74v2 Set | 160123_at | - | - |
| Murine Genome U74v2 Set | 160123_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427903_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427903_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427903_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427903_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7050537 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2626143 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2626143 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2626143 | - | - |

Phpt1
Genomic Map