Assay Details
| Gene Symbol: Ptgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008963.2 | NP_032989.2 | 2 - 3 | 327 | 75 | |
| GenBank mRNA | AB006361.1 | - | 2 - 3 | 310 | 75 | |
| AK002382.1 | - | 1 - 2 | 152 | 75 | ||
| AK131859.1 | - | 2 - 3 | 327 | 75 | ||
| BC038083.1 | - | 1 - 2 | 403 | 4481308 | 75 | |
| BC043015.1 | - | 2 - 3 | 313 | 5683706 | 75 | |
| X89222.1 | - | 2 - 3 | 298 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptgds
Genomic Map