Assay Details
| Gene Symbol: Clic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027085.3 | NP_081361.1 | 5 - 6 | 588 | 63 | |
| GenBank mRNA | AK009020.1 | - | 5 - 6 | 590 | 63 | |
| BC060967.1 | - | 5 - 6 | 593 | 30297308 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979508 | NM_027085.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979508 | AK009020.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979508 | BC060967.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665438 | AK081117.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665438 | AK147993.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665438 | BC115689.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665438 | BC115690.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665438 | XM_001474974.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665438 | XM_001476637.1 | refseq_mrna_mouse_XM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140647 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979508 | NM_027085.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979508 | AK009020.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979508 | BC060967.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P236755 | NM_027085.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P236755 | AK009020.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P236755 | BC060967.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072277 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140647 | - | - |
| Whole Mouse Genome G4122A | A_51_P236755 | NM_027085.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P236755 | AK009020.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P236755 | BC060967.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125433 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140647 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02001932 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02001930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02001917 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02001926 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02001973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02001960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02001917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02001946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02001966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02001979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02001902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02001912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02001932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02001976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01791836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02052324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02001973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02001942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02001958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072269 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10470105 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470106 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470108 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470107 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429574_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10470101 | - | - |
| Mouse Genome 430 2.0 Array | 1429574_at | - | - |
| Mouse Genome 430 2.0 Array | 1429574_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69454_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69454_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200002515.mm.1 | NM_027085.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002515.mm.1 | NM_027085.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002515.mm.1 | TC0200000419.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200002513.mm.1 | NM_027085.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002513.mm.1 | NM_027085.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002513.mm.1 | TC0200000419.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429574_PM_at | - | - |
| Mouse Expression Set 430 | 1429574_at | - | - |
| Murine Genome U74v2 Set | 160884_at | - | - |
| Murine Genome U74v2 Set | 160884_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429574_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429574_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429574_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429574_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 423725 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4060670 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4060670 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004060670 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2650667 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2650667 | - | - |
| mouse-6 v1.1 (Illumina) | scl21176.6.1_21-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4060670 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21176.6.1_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060670 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2650667 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2650667 | - | - |

Clic3
Genomic Map