Assay Details
| Gene Symbol: Qsox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042036.1 | - | 1 - 2 | 231 | 77 | |
| AK140324.1 | - | 4 - 5 | 609 | 77 | ||
| AK157992.1 | - | 4 - 5 | 602 | 77 | ||
| AK165642.1 | - | 4 - 5 | 611 | 77 | ||
| BC030934.1 | - | 4 - 5 | 643 | 4920915 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Qsox2
Genomic Map