Assay Details
| Gene Symbol: Ubac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133835.2 | NP_598596.2 | 5 - 6 | 768 | 70 | |
| GenBank mRNA | AK157857.1 | - | 5 - 6 | 602 | 70 | |
| AK217076.1 | - | 1 - 2 | 86 | 70 | ||
| BC021811.1 | - | 5 - 6 | 719 | 3662474 | 70 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ubac1
Genomic Map