Assay Details
| Gene Symbol: Ubac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133835.2 | NP_598596.2 | 7 - 8 | 1118 | 100 | |
| GenBank mRNA | AK004060.1 | - | 1 - 2 | 206 | 100 | |
| AK157857.1 | - | 7 - 8 | 952 | 100 | ||
| AK161969.1 | - | 2 - 3 | 321 | 100 | ||
| AK197619.1 | - | 1 - 2 | 70 | 100 | ||
| AK209158.1 | - | 2 - 3 | 294 | 100 | ||
| AK209934.1 | - | 1 - 2 | 70 | 100 | ||
| AK213285.1 | - | 2 - 3 | 291 | 100 | ||
| BC021811.1 | - | 7 - 8 | 1069 | 3662474 | 100 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ubac1
Genomic Map