Assay Details
| Gene Symbol: Ubac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133835.2 | NP_598596.2 | 8 - 9 | 1197 | 91 | |
| GenBank mRNA | AK157857.1 | - | 8 - 9 | 1031 | 91 | |
| BC021811.1 | - | 8 - 9 | 1148 | 3662474 | 91 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ubac1
Genomic Map