Assay Details
| Gene Symbol: Inpp5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290437.1 | NP_001277366.1 | 9 - 10 | 2019 | 64 | |
| NM_033134.3 | NP_149125.1 | 8 - 9 | 2303 | 64 | ||
| NR_110957.1 | 8 - 9 | 2390 | 64 | |||
| GenBank mRNA | AF226683.1 | - | 8 - 9 | 2265 | 64 | |
| AK047245.1 | - | 6 - 7 | 2232 | 64 | ||
| AK080075.1 | - | 8 - 9 | 2285 | 64 | ||
| AK154097.1 | - | 8 - 9 | 2304 | 64 | ||
| AK158135.1 | - | 8 - 9 | 2288 | 64 | ||
| AK170174.1 | - | 8 - 9 | 2373 | 64 | ||
| AK170786.1 | - | 8 - 9 | 2260 | 64 | ||
| AK170812.1 | - | 7 - 8 | 2338 | 64 | ||
| BC052717.1 | - | 8 - 9 | 2272 | 6837339 | 64 | |
| BC057566.1 | - | 8 - 9 | 2355 | 64 | ||
| BC080295.1 | - | 8 - 9 | 2268 | 30354397 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inpp5e
Genomic Map