Assay Details
| Gene Symbol: Vrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011705.3 | NP_035835.1 | 13 - 14 | 1327 | 98 | |
| GenBank mRNA | AF080253.1 | - | 13 - 14 | 1281 | 98 | |
| AK088674.1 | - | 13 - 14 | 1310 | 98 | ||
| BC051072.1 | - | 12 - 13 | 2261 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vrk1
Genomic Map