Assay Details
| Gene Symbol: Ppp6r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172894.2 | NP_766482.2 | 15 - 16 | 2228 | 93 | |
| GenBank mRNA | AK122450.1 | - | 11 - 12 | 3331 | 93 | |
| BC053076.1 | - | 15 - 16 | 2228 | 5698212 | 93 | |
| BC141039.1 | - | 14 - 15 | 1775 | 9055651 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp6r1
Genomic Map